3-Cyanobenzamide 96%

Supplier: Apollo Scientific

APOSOR946455-10GEA 967 EUR
APOSOR946455-10G APOSOR946455-1G APOSOR946455-5G
3-Cyanobenzamide 96%
3-Cyanobenzamide
Formula: C₈H₆N₂O
MW: 146,1460 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017574
CAS Number: 3441-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR