tert-Butyl trichloroacetimidate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
tert-Butyl 2,2,2-trichloroacetimidate
, t-butyl-2,2,2-trichloroacetimidate
, t-Butyl trichioroacetimidate
APOSOR9573-1GEA
36.4
EUR
APOSOR9573-1G
APOSOR9573-5G
APOSOR9573-25G
tert-Butyl trichloroacetimidate 98%
tert-Butyl trichloroacetimidate
Formula:
C₆H₁₀Cl₃NO MW: 218,502 g/mol Boiling Pt: 64…66 °C/15mmHg Melting Pt: 20…22 °C Density: 1,222 g/cm³ (25 °C) Flash Pt: 55 °C Storage Temperature: Freezer |
MDL Number:
MFCD00077410 CAS Number: 98946-18-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...