4-Amino-3-(trifluoromethoxy)benzonitrile 97%

Supplier: Apollo Scientific
Warning

APOSPC1100BF-10GEA 178 EUR
APOSPC1100BF-10G
4-Amino-3-(trifluoromethoxy)benzonitrile 97%
4-Amino-3-(trifluoromethoxy)benzonitrile
Formula: C₈H₅F₃N₂O
MW: 202,1300 g/mol
Melting Pt: 71…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00204177
CAS Number: 175278-23-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR