α,2-Dibromo-4-fluorotoluene

Supplier: Apollo Scientific
Danger

Synonyms: 2-Bromo-4-fluorobenzyl bromide

APOSPC1236-1GEA 12.6 EUR
APOSPC1236-1G
α,2-Dibromo-4-fluorotoluene
α,2-Dibromo-4-fluorotoluene
Formula: C₇H₅Br₂F
MW: 267,92
Boiling Pt: 128…130 °C/23mmHg
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD03840524
CAS Number: 61150-57-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR