Potassium-3-hydroxyphenyltrifluoroborate

Supplier: Apollo Scientific

Synonyms: Potassium trifluoro(3-hydroxyphenyl)borate

APOSPC200425-1GEA 56.8 EUR
APOSPC200425-1G
Potassium-3-hydroxyphenyltrifluoroborate
Potassium 3-hydroxyphenyltrifluoroborate
Formula: C₆H₅BF₃KO
MW: 200,0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784422
CAS Number: 871231-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR