4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV) 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4'-Butyl-2,2,2-trifluoroacetophenone1-(4-Butylphenyl)-2,2,2-trifluoroethanone

APOSPC2499-1GEA 408 EUR
APOSPC2499-1G
4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV) 97%
4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV)
Formula: C₁₂H₁₃F₃O
MW: 230,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00214157
CAS Number: 40739-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR