4-Fluorodiphenylamine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Fluoro-N-phenylaniline
APOSPC2801-1GEA
132
EUR
APOSPC2801-1G
4-Fluorodiphenylamine 98%
4-Fluorodiphenylamine
Formula:
C₁₂H₁₀FN MW: 187,2129 g/mol Boiling Pt: 176…177 °C/15mmHg Melting Pt: 31…34 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318538 CAS Number: 330-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...