4-(Ethoxycarbonyl)-3-fluorophenylboronic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Ethoxycarbonyl-3-fluorophenylboronic acid4-Ethoxycarbonyl-3-fluorobenzeneboronic acid

APOSPC2999-1GEA 46.6 EUR
APOSPC2999-1G
4-(Ethoxycarbonyl)-3-fluorophenylboronic acid 95%
4-(Ethoxycarbonyl)-3-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
MW: 211,98 g/mol
Melting Pt: 158…161 °C
Storage Temperature: Ambient
MDL Number: MFCD06656274
CAS Number: 874288-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR