5-Fluoro-2-(trifluoromethyl)phenylboronic acid
Supplier: Apollo Scientific
|
Warning
|
APOSPC3019-500MGEA
16.1
EUR
APOSPC3019-500MG
5-Fluoro-2-(trifluoromethyl)phenylboronic acid
5-Fluoro-2-(trifluoromethyl)phenylboronic acid
Formula:
C₇H₅BF₄O₂ MW: 207,9180 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06656275 CAS Number: 928053-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...