4-Chloro-2-fluoro-3-methoxybenzylamine 95%
Supplier: Apollo Scientific
|
Danger
|
APOSPC302861-1GEA
147
EUR
APOSPC302861-1G
4-Chloro-2-fluoro-3-methoxybenzylamine 95%
4-Chloro-2-fluoro-3-methoxybenzylamine
Formula:
C₈H₉ClFNO MW: 189,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687209 CAS Number: 1323966-34-2 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...