3-Fluoro-5-(trifluoromethyl)benzophenone 97%
Supplier: Apollo Scientific
|
Warning
|
APOSPC4374J-1GEA
36.4
EUR
APOSPC4374J-1G
3-Fluoro-5-(trifluoromethyl)benzophenone 97%
3-Fluoro-5-(trifluoromethyl)benzophenone
Formula:
C₁₄H₈F₄O Storage Temperature: Ambient |
MDL Number:
MFCD00061256 CAS Number: 239087-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...