3,5-Difluoro-4-formylphenylboronic acid
Supplier: Apollo Scientific
Synonyms:
(3,5-Difluoro-4-formylphenyl)boronic acid
APOSPC448009-1GEA
77.1
EUR
APOSPC448009-1G
3,5-Difluoro-4-formylphenylboronic acid
3,5-Difluoro-4-formylphenylboronic acid
Formula:
C₇H₅BF₂O₃ MW: 185,9206 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07784344 CAS Number: 870718-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...