2-Chloro-3-fluorobenzaldehyde 98%

Supplier: Apollo Scientific

APOSPC499011-5GEA 30.1 EUR
APOSPC499011-100G APOSPC499011-5G APOSPC499011-25G
2-Chloro-3-fluorobenzaldehyde 98%
2-Chloro-3-fluorobenzaldehyde
Formula: C₇H₄ClFO
Melting Pt: 29…32 °C
Storage Temperature: Ambient
MDL Number: MFCD07782053
CAS Number: 96516-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR