5-(2-Fluorophenyl)-3-pyridinecarbaldehyde
Supplier: Apollo Scientific
APOSPC510093-5GEA
1620
EUR
APOSPC510093-5G
5-(2-Fluorophenyl)-3-pyridinecarbaldehyde
5-(2-Fluorophenyl)-3-pyridinecarbaldehyde
Formula:
C₁₂H₈FNO Storage Temperature: Ambient |
CAS Number:
887973-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...