4-Fluoro-3-methylbenzenesulphonamide 98%
Supplier: Apollo Scientific
|
Warning
|
APOSPC5224-1GEA
46.6
EUR
APOSPC5224-1G
4-Fluoro-3-methylbenzenesulphonamide 98%
4-Fluoro-3-methylbenzenesulphonamide
Formula:
C₇H₈FNO₂S Storage Temperature: Ambient |
MDL Number:
MFCD01365826 CAS Number: 379254-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...