1,1,2-Trifluordecene 97%

Supplier: Apollo Scientific
Warning

APOSPC6459-25GEA 248 EUR
APOSPC6459-25G
1,1,2-Trifluordecene 97%
1,1,2-Trifluordecene
Formula: C₁₀H₁₇F₃
Boiling Pt: 91 °C/90mmHg
Storage Temperature: Ambient
MDL Number: MFCD00156047
CAS Number: 692-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR