2-Bromoheptafluoronaphthalene 85%

Supplier: Apollo Scientific
Warning

PC7930-250MG
APOSPC7930-250MGEA 104 EUR
APOSPC7930-250MG
2-Bromoheptafluoronaphthalene 85%
2-Bromoheptafluoronaphthalene
Formula: C₁₀BrF₇
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD07784218
CAS Number: 27041-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR