3,5-Bis(trifluoromethyl)styrene 98%
Supplier: Apollo Scientific
|
Warning
|
APOSPC9182-1GEA
53.9
EUR
APOSPC9182-1G
3,5-Bis(trifluoromethyl)styrene 98%
3,5-Bis(trifluoromethyl)styrene
Formula:
C₁₀H₆F₆ MW: 240,15 g/mol Boiling Pt: 59…61 °C/20mmHg Melting Pt: 44415 °C Density: 1,334 g/cm³ (25 °C) Flash Pt: 50 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075538 CAS Number: 349-59-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...