TRIS-HEPES-SDS Running Buffer
Supplier: Apollo
APOSBI2808-10PACKSEA 0 EUR
APOSBI2808-10PACKS APOSBI2808-5PACKS
TRIS-HEPES-SDS Running Buffer
Buffers Electrophoresis Buffers Running Buffers
Pre-weighed sachet containing sufficient buffer salts to create 1 litre of solution containing 1M TRIS, 1M HEPES and 30mM SDS.