3,3',5,5'-Tetramethylbenzidine (TMB) Liquid Chromogenic Substrate
|
Warning
|
Synonyms:
TMB
, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
TMB Ready-To-Use chromogenic substrate contains 3,3',5,5'- tetramethylbenzidine and H202 in a Citric Acid buffer and is recommended for use in ELISA procedures. TMB is a substrate for horseradish peroxidase. It produces a soluble blue color product that can be read spectrophotometrically at 370 or 620-650 nm. The reaction may be amplified threefold when stopp ed with acid, generating a yellow color with its absorption maximum at 450nm. After stopping, it should be read within 30 minutes. Not recommended for use in immunohistochemical or blotting assays. Stability: Minimum of 1 year if stored at 2-8 °C.
Formula:
C₁₆H₂₀N₂ MW: 240,35 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00007748 CAS Number: 54827-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...