VariFlex
Supplier: Agilent Technologies
AGLS240105EA 421 EUR
AGLS240105
VariFlex
Protein Expression Protein Expression Systems
The VariFlex protein expression system provides vectors with unique tags that enhance the utility of E. coli as an expression host.
  • All three SET tags in every vector set
  • Gentle SBP purification tag generates high-yield, pure proteins
Delivery information: Includes tag offering are three different SET tags which increase protein solubility and the SBP purification tag for high-yield, pure proteins.
Caution: For research use only. Not for use in diagnostic procedures.