S-Nitrosoglutathione (GSNO)

Supplier: BioVision

Synonyms: S-Nitroso-L-GlutathioneGSNO

BIOV1580-100EA 350 EUR
BIOV1580-100 BIOV1580-25
S-Nitrosoglutathione (GSNO)
S-Nitrosoglutathione (GSNO)
Formula: C₁₀H₁₆N₄O₇S
MW: 336,319 g/mol
CAS Number: 57564-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR