You Searched For: Soldering Tips
Compatible with pencils and irons, the interchangeable soldering tips come in wide variety of styles and sizes to accommodate any specific procedure. The pieces also serve as replacement parts for corrosion affected points that will no longer melt the fusing metal. Manufactured with temperature resistant iron, the bits can be removed from the wand by unscrewing or pushing in and pulling. Proper sponge cleaning will prolong the life of the heat transferring copper in soldering tips.
1 results were found
Sort Results
List View
Easy View