ID-8 ≥98% (by TLC)

Supplier: BioVision
Warning

Synonyms: 1-(4-Methoxyphenyl)-2-methyl-3-nitro-1H-indol-6-ol

BIOV2562-25EA 558 EUR
BIOV2562-25 BIOV2562-5
ID-8 ≥98% (by TLC)
ID-8

ID-8 is a cell culture supplement that can sustain self-renewal and pluripotency of mouse embryonic stem cells in vitro


At 10 μM it stimulates proliferation in serum-free media at a steady rate for more than 30 days. Also inhibits dual-specificity tyrosine phosphorylation-regulated kinase (DYRK).


Caution: FOR RESEARCH USE ONLY! Not to be used in humans.

Formula: C₁₆H₁₄N₂O₄
MW: 298,30 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00593367
CAS Number: 147591-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR