EZClick™ palmitoylated protein assay kit (FACS/microscopy), red fluorescence
Supplier: BioVision
BIOVK416-100EA 0 EUR
BIOVK416-100
EZClick™ palmitoylated protein assay kit (FACS/microscopy), red fluorescence
For labeling and detection of palmitoylated proteins.
  • Highly specific
  • Simple procedure
  • Robust method for labeling and detection of palmitoylated proteins
Palmitoylation occurs when fatty acids, such as palmitic acid are covalently attached to the side chains of cysteine (and less frequently to serine, threonine side chains) in proteins. This type of post-translational modification greatly affects cellular trafficking, compartmentalization, and membrane tethering. Palmityl transferase (PAT), the enzyme responsible for this modification, transfers a palmitate moiety from palmitoyl-CoA to the thiol group of cysteine in the target protein. Compared to myristylation, palmitoylation is reversible and the reverse reaction is catalysed by thioesterases. The Palmitoylation/DE palmitoylation cycle plays an important role when modified protein shuttles between cellular compartments. PAT mutations are associated with many neurological diseases and cancer progression. BioVision offers EZClick™ palmitoylated protein assay kit, a highly specific, simple and robust method for labelling and detection of palmitoylated proteins. The kit uses a modified palmitic acid that is fed directly into the cells and gets incorporated into proteins during or post translation. This post translational modification can be followed by click reaction with an azide-containing dye. The assay kit offers a powerful method for imaging localization, trafficking, and dynamics of Palmitoylated proteins, or detection by FACS for quantitative studies. Labelled palmitoylated proteins can be directly detected in 1D or 2D gels using the appropriate excitation sources, or enriched by immunoprecipitation with biotin-azide or antibodies prior to proteomic analysis. We provide sufficient materials for 100 assays in a 96-well plate format.