Yeast poly(A) polymerase
Supplier: BioVision
M1215-100EA 237 EUR
M1215-100
Yeast poly(A) polymerase
Enzymes
Poly(A) polymerase catalyses the template independent addition of adenosine residues onto the 3' ends of polyribonucleotides.
  • Labelling of RNA with ATP or cordycepin
  • Poly(A) tailing of RNA for cloning or affinity purification
  • Increasing translation of RNA transferred into eukaryotic cells