Troponin I (Rat) ELISA kit
Supplier: BioVision
BIOVE4737-100EA 0 EUR
BIOVE4737-100
Troponin I (Rat) ELISA kit
A sandwich ELISA kit for in vitro quantitative determination of rat Troponin I concentrations in serum, plasma and other biological fluids.
  • 96 assays
  • Species Reactivity: Rat
  • Coefficient of variation is <10%
Delivery information: Kit components: Biotinylated detection Ab (100×), stop solution, wash buffer (25×), biotinylated detection antibody diluent, micro ELISA plate, HRP conjugate (100×), HRP conjugate diluent, standard and sample diluent, substrate reagent, standard and plate sealer.
Caution: For research use only! Not for use in humans. This product can expose you to chemicals including TMB, which is [are] known to the State of California to cause cancer.