ATPase activity assay kit (Colorimetric)
Supplier: BioVision
BIOVK417-100EA 0 EUR
BIOVK417-100
ATPase activity assay kit (Colorimetric)
Quick and easy method for monitoring ATPase activity in various samples.
  • Simple procedure; takes less than 1 hour
  • Fast and convenient
ATPase (Adenosine Triphosphatase: EC 3.6.1.3) is an important enzyme for maintaining the cell membrane potential, transporting ions and regulating cellular volume. It catalyses the decomposition of ATP into ADP and a free phosphate ion. The hydrolysis of ATP is highly exergonic releasing energy that is utilized in several cellular processes. There are many classes of ATPases including Na+/K+ -ATPase, H+/K+ -ATPase, Ca2+ -ATPase, etc. The deficiency of mitochondrial ATPase is serious: for example, Na+/K+-ATPase deficiency increases anxiety-related behaviour, while Ca2+ -ATPase deficiency leads to exertional muscle pain syndrome. Therefore, accurate detection of ATPase activity is valuable for the diagnosis and mechanistic studies of some of these diseases. BioVision’s ATPase Activity Assay kit provides a quick and easy method for monitoring ATPase activity in various samples. In the assay, ATPase hydrolyses ATP releasing ADP and a free phosphate ion, and through linked reactions, a strong, stable chromophore is generated (OD 650 nm). The assay is simple, sensitive, high -throughput adaptable and can detect ATPase Activity less than 0,005 U/L