Methylglyoxal assay kit (colorimetric)
Supplier: BioVision
BIOVK500-100EA 440 EUR
BIOVK500-100
Methylglyoxal assay kit (colorimetric)
First kit in the market that can detect Methylglyoxal in samples.
  • 100 assays
  • Species reactivity- Eukaryotes
  • Simple procedure; takes ~ 2 hours
  • Fast and convenient
  • The assay is sensitive, stable and high-throughput adaptable
  • Useful for the estimation of Methylglyoxal in biological samples
This Kit provides a simple in vitro assay for detecting Methylglyoxal levels in biological samples.
Delivery information: Kit components: MG assay buffer, substrate mix A, substrate mix B, enzyme mix A, enzyme mix B, enzyme mix C and MG standard (20 mM).
Caution: For research use only! Not for use in humans.