Insect genomic DNA kit
Supplier: BioVision
K1412-250EA 530 EUR
K1412-250
Insect genomic DNA kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
For efficient recovery of genomic DNA up to 60 kb in size from insects, arthropods.
  • For efficient recovery of genomic DNA up to 60 kb in size from insects, arthropods, and some plant tissue samples rich in polysaccharides.
For efficient recovery of genomic DNA up to 60 kb in size from insects, arthropods, and some plant tissue samples rich in polysaccharides.