RNase R
Supplier: BioVision
M1228-500EA 327 EUR
M1228-500
RNase R
Enzymes
An E. coli exoribonuclease
  • Simple procedure
  • Fast and convenient
RNase R is an E. coli exoribonuclease which exhibits 3’-to-5’ exonuclease activity, efficiently digesting nearly all linear RNA species.