Coronavirus recombinant SARS-CoV-2 3C-like proteinase (fromE. coli)
Supplier: BioVision
BIOVP1606-50EA 0 EUR
BIOVP1606-50
Coronavirus recombinant SARS-CoV-2 3C-like proteinase (fromE. coli)
SARS-CoV-2 is a novel strain of coronavirus which causes the infectious disease COVID-19. 3C‐like protease is the main cysteine protease of the virus which hydrolyses viral polyproteins to produce functional proteins which is essential for coronavirus replication. It is an important therapeutic target for COVID-19.