3M™ instant microbe removing wipes are cleaning wipes that trap, hold, and remove microbes to virtually eliminate cross contamination. They work using just water, removing microbes instantly. Ideal for high-touch, fast-turnaround areas.
- Quickly traps, holds and removes 99,9% of microbes including SARS-Cov-2* to virtually eliminate cross contamination
- Works using just water; no disinfectant, no specific PPE required
- Easy to use; works with no dwell time
- Also works with quat disinfectants
- Refillable containers reduce waste; if wipes dry out just add water
- *based on 3rd party testing with water only
Trap, hold, and remove microbes with 3M™ Instant Microbe Removing Wipes. These wipes remove microbes from surfaces instantly using just water, trapping them in the wipe to virtually eliminate cross contamination. They’re easy to use, with no dwell time, making them ideal for high-touch, fast-turnaround areas. They’re also designed to work with quat disinfectants.