Glecaprevir ≥98%

Supplier: BioVision

Synonyms: (33R,35S,91R,92R,5S,E)-5-(tert-butyl)-N-((1R,2R)-2-(difluoromethyl)-1-(((1-methylcyclopropyl)sulfonyl)carbamoyl)cyclopropyl)-14,14-difluoro-4,7-dioxo-2,8,10-trioxa-6-aza-1(2,3)-quinoxalina-3(3,1)-pyrrolidina-9(1,2)-cyclopentanacyclotetradecABT-493ABT493

B2347-5EA 186 EUR
B2347-5
Glecaprevir ≥98%
Glecaprevir

Glecaprevir is a hepatitis C virus nonstructural protein 3/4A protease inhibitor that targets the viral RNA replication.


  • A Hepatitis C Virus NS3/4A Protease Inhibitor

Formula: C₃₈H₄₆F₄N₆O₉S
Storage Temperature: Freezer
CAS Number: 1365970-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR