DTSSP (3,3'-Dithiobis(sulphosuccinimidyl propionate))
Supplier: G Biosciences
Synonyms:
DTSSP
BC08EA
612
EUR
BC08
DTSSP (3,3'-Dithiobis(sulphosuccinimidyl propionate))
DTSSP (3,3'-Dithiobis(sulphosuccinimidyl propionate))
3,3’-Dithiobis(sulfosuccinimidyl propionate) Homobifunctional Cross Linker.
Formula:
C₁₄H₁₆N₂O₁₄S₄ MW: 564,542 g/mol |
MDL Number:
MFCD01861949 CAS Number: 81069-02-5 |
Specification Test Results
Spacer Arm (Å) | 12 |
Reactive Toward | Primary Amines |
Membrane Permeable | NO |
Water Soluble | YES |
Cleavable/ Reversible | Reducing Agents (Thiols) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...