1-Oxa-9-azaspiro[5.5]undecane hydrochloride 95%
|
|
BLDPBD00751780-250EA
129
EUR
BLDPBD00751780-250
BLDPBD00751780-1G
BLDPBD00751780-100
1-Oxa-9-azaspiro[5.5]undecane hydrochloride 95%
1-Oxa-9-azaspiro[5.5]undecane hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₉H₁₇NO·ClH MW: 191,698 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11036126 CAS Number: 1414958-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...