1,2:5,6-Diepoxyhexane 97%
|
|
Synonyms:
1,5-Hexadiene diepoxide
BD00768047-1G
BD00768047-5G
BD00768047-25G
BLDPBD00768047-1GEA
24.2
EUR
BLDPBD00768047-1G
BLDPBD00768047-5G
BLDPBD00768047-25G
1,2:5,6-Diepoxyhexane 97%
1,2:5,6-Diepoxyhexane
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₁₀O₂ MW: 114,1424 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01723447 CAS Number: 1888-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...