3-Cyclohexyl-DL-alanine 97%

Synonyms: 2-Amino-3-cyclohexylpropanoic acid

BLDPBD00772587-1GEA 26.3 EUR
BLDPBD00772587-1G BLDPBD00772587-250 BLDPBD00772587-5G
3-Cyclohexyl-DL-alanine 97%
3-Cyclohexyl-DL-alanine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₉H₁₇NO₂
MW: 171,237 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00996128
CAS Number: 4441-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR