(S)-AntPhos 97%

Synonyms: (S)-4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

BLDPBD00943764-250EA 171 EUR
BLDPBD00943764-250 BLDPBD00943764-100 BLDPBD00943764-1G
(S)-AntPhos 97%
(S)-AntPhos

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₅H₂₃OP
MW: 370,423 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31630706
CAS Number: 1807740-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR