1-Dodecyl-3-methylimidazolium bromide 98%

Synonyms: 1-Dodecyl-3-methylimidazolium Bromide

BLDPBD00958250-100EA 118 EUR
BLDPBD00958250-100 BLDPBD00958250-5G BLDPBD00958250-25G
1-Dodecyl-3-methylimidazolium bromide 98%
1-Dodecyl-3-methylimidazolium bromide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₆H₃₁BrN₂
MW: 331,3347 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17014729
CAS Number: 61546-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR