2-Fluoro-5-(methylcarbamoyl)phenylboronic acid 98%
|
|
Synonyms:
2-Fluoro-5-(methylcarbamoyl)benzeneboronic acid
, (2-Fluoro-5-(methylcarbamoyl)phenyl)boronic acid
BLDPBD01088552-250EA
30.5
EUR
BLDPBD01088552-250
BLDPBD01088552-1G
BLDPBD01088552-5G
2-Fluoro-5-(methylcarbamoyl)phenylboronic acid 98%
2-Fluoro-5-(methylcarbamoyl)phenylboronic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₈H₉BFNO₃ MW: 196,97 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08436028 CAS Number: 874289-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...