1,5-Dioxa-9-azaspiro[5.5]undecane hydrochloride 97%

BLDPBD01089038-5GEA 473 EUR
BLDPBD01089038-5G BLDPBD01089038-250 BLDPBD01089038-1G
1,5-Dioxa-9-azaspiro[5.5]undecane hydrochloride 97%
1,5-Dioxa-9-azaspiro[5.5]undecane hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₁₆ClNO₂
MW: 193,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036126
CAS Number: 71879-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR