4-Chloro-3-oxobutanoic acid 97%

BLDPBD01095908-100EA 27.3 EUR
BLDPBD01095908-100 BLDPBD01095908-1G BLDPBD01095908-5G BLDPBD01095908-250
4-Chloro-3-oxobutanoic acid 97%
4-Chloro-3-oxobutanoic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₄H₅ClO₃
MW: 136,534 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00129128
CAS Number: 27807-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR