2-Chloro-3-(trifluoromethyl)phenylboronic acid 98+%

BD01390252-1G BD01390252-25G BD01390252-5G
BLDPBD01390252-1GEA 18.9 EUR
BLDPBD01390252-1G BLDPBD01390252-25G BLDPBD01390252-5G
2-Chloro-3-(trifluoromethyl)phenylboronic acid 98+%
2-Chloro-3-(trifluoromethyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₅BClF₃O₂
MW: 224,3726 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258745
CAS Number: 957061-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR