2-Bromo-6-hydroxybenzonitrile 99%

Synonyms: 3-Bromo-2-cyanophenol

BLDPBD01391661-1GEA 44.1 EUR
BLDPBD01391661-1G BLDPBD01391661-100 BLDPBD01391661-5G BLDPBD01391661-25G
2-Bromo-6-hydroxybenzonitrile 99%
2-Bromo-6-hydroxybenzonitrile

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₄BrNO
MW: 198,017 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12828440
CAS Number: 73289-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR