4-Aminophenylboronic acid hydrochloride 98%

Synonyms: 4-Aminobenzeneboronic acid hydrochloride,(4-Aminophenyl)boronic acid hydrochloride

BD01405023-1G BD01405023-5G BD01405023-25G BD01405023-100
BLDPBD01405023-1GEA 8.4 EUR
BLDPBD01405023-1G BLDPBD01405023-5G BLDPBD01405023-25G BLDPBD01405023-100
4-Aminophenylboronic acid hydrochloride 98%
4-Aminophenylboronic acid hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₉BClNO₂
MW: 173,41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001333
CAS Number: 80460-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR