5-Amino-2,4,6-triiodoisophthalic acid 98+%

Supplier: BLD PHARMATECH GMBH

BLDPBD01414217-25GEA 18.9 EUR
BLDPBD01414217-25G BLDPBD01414217-5G
5-Amino-2,4,6-triiodoisophthalic acid 98+%
5-Amino-2,4,6-triiodoisophthalic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₄I₃NO₄
MW: 558,84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190167
CAS Number: 35453-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR