(S)-1-Boc-3-hydroxypiperidine 98%

Synonyms: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate(3S)-3-Hydroxypiperidine, N-BOC protected

BLDPBD0325-1GEA 6.3 EUR
BLDPBD0325-1G BLDPBD0325-0.5G BLDPBD0325-25G BLDPBD0325-5G BLDPBD0325-100G BLDPBD0325-10G BLDPBD0325-500G
(S)-1-Boc-3-hydroxypiperidine 98%
(S)-1-Boc-3-hydroxypiperidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₁₉NO₃
MW: 201,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115307
CAS Number: 143900-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR