Ethyl-2,4-dimethylbenzoate 98%

Synonyms: Ethyl 2,4-dimethylbenzoate

BLDPBD0597-5GEA 49.4 EUR
BLDPBD0597-5G BLDPBD0597-25G BLDPBD0597-100G
Ethyl-2,4-dimethylbenzoate 98%
Ethyl 2,4-dimethylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015438
CAS Number: 33499-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR