2-Amino-4-phenylphenol 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 3-Amino-4-hydroxybiphenyl3-Amino-[1,1'-biphenyl]-4-ol

BLDPBD0762-5GEA 133 EUR
BLDPBD0762-5G BLDPBD0762-1G
2-Amino-4-phenylphenol 98%
2-Amino-4-phenylphenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₂H₁₁NO
MW: 185,2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059187
CAS Number: 1134-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR